Invitrogen
Manufacturer Code:PA542969
Catalog # PIPA542969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EDEM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ER degradation enhancer mannosidase alpha-like 1 EDEM KIAA0212; ER degradation enhancer, mannosidase alpha-like 1; ER degradation ER degradation enhancer, mannosidase alpha-like 1; ER degradation-enhancing alpha-mannosidase-like 1; ER degradation-enhancing alpha-mannosidase-like protein 1
Gene Aliases: A130059K23Rik; EDEM; EDEM1; KIAA0212; mKIAA0212; RGD1563633
UniProt ID: (Human) Q92611, (Mouse) Q925U4
Entrez Gene ID: (Human) 9695, (Bovine) 511008, (Rabbit) 100346313, (Dog) 476547, (Mouse) 192193, (Rat) 297504, (Guinea pig) 100714413
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.