Invitrogen
Manufacturer Code:PA559237
Catalog # PIPA559237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human EDC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: enhancer of mRNA decapping 3 homolog; Enhancer of mRNA-decapping protein 3; hYjeF_N2; hYjeF_N2-15q23; LSM16 homolog; LSM16 homolog (EDC3, S. cerevisiae); PP844 LSM16 YJDC YJEFN2 LSM16 homolog (EDC3 S. cerevisiae) enhancer of mRNA-decapping protein 3 hYjeF&lowbarN2 hYjeF&lowbarN2-15q23 yjeF N-terminal domain-containing protein 2 yjeF domain containing yjeF domain-containing protein 1 yjeF&lowbarN2; yjeF domain containing; YjeF domain-containing protein 1; YjeF N-terminal domain-containing protein 2; YjeF_N2
Gene Aliases: EDC3; LSM16; MRT50; PP844; YJDC; YJEFN2
UniProt ID: (Human) Q96F86
Entrez Gene ID: (Human) 80153
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.