Invitrogen
Manufacturer Code:PA5116422
Catalog # PIPA5116422
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 162-197 amino acids from the Central region of (DANRE) edc3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA517853 EDC3 enhancer of mRNA decapping 3 enhancer of mRNA decapping 3 homolog enhancer of mRNA decapping 3 homolog (S. cerevisiae) enhancer of mRNA-decapping protein 3 hYjeF_N2 hYjeF_N2-15q23 LSM16 LSM16 homolog LSM16 homolog (EDC3 S. cerevisiae) MRT50 PP844 RGD1306898 YJDC yjeF domain containing yjeF domain-containing protein 1 YjeF N-terminal domain-containing protein 2 yjeF_N2 YJEFN2; LSM16 homolog (EDC3, S. cerevisiae); LSM16 protein homolog; yjeF domain containing; yjeF domain-containing protein 1
Gene Aliases: im:7151288; lsm16; wu:fb82d07; yjdc; zgc:112006
Entrez Gene ID: (Zebrafish) 504054
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.