Invitrogen
Manufacturer Code:PA542588
Catalog # PIPA542588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human EDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ectodermal dysplasia 1, anhidrotic; Ectodermal dysplasia protein; Ectodysplasin 1, isoform A2; Ectodysplasin A ED1 ED1-A1 ED1-A2 EDA1 EDA2 HED XHED XLHED ODT1 STHAGX1; Ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; EDA protein; EDA protein homolog; nackt; nkt; oligodontia 1; Tabby protein; tumor necrosis factor ligand 7C; X-linked anhidroitic ectodermal dysplasia protein
Gene Aliases: ECTD1; ED1; ED1-A1; ED1-A2; EDA; EDA-A1; EDA-A2; EDA1; EDA2; HED; HED1; ODT1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; TNLG7C; XHED; XLHED
UniProt ID: (Human) Q92838, (Bovine) Q9BEG5, (Mouse) O54693
Entrez Gene ID: (Human) 1896, (Goat) 100861240, (Rabbit) 100340021, (Dog) 491935, (Bovine) 616179, (Horse) 100065499, (Rat) 302424, (Guinea pig) 100734442, (Mouse) 13607, (Zebrafish) 798740
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.