Proteintech
Manufacturer Code:213761AP150UL
Catalog # 50-173-3393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ECM2 Fusion Protein Ag15913 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECM2 Extracellular matrix protein 2 Matrix glycoprotein SC1 Matrix glycoprotein SC1/ECM2; Extracellular matrix protein 2; extracellular matrix protein 2, female organ and adipocyte specific; extracellular matrix protein 2-like; Matrix glycoprotein SC1/ECM2; Tenonectin
Gene Aliases: 9030618O22Rik; BC065151; ECM2; tenonectin
UniProt ID: (Human) O94769, (Mouse) Q5FW85
Entrez Gene ID: (Human) 1842, (Mouse) 407800, (Rat) 100361383
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.