Invitrogen
Manufacturer Code:PA542882
Catalog # PIPA542882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ECM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Extracellular matrix protein 1; Secretory component p85; testicular tissue protein Li 61
Gene Aliases: AI663821; ECM1; p85; URBWD
UniProt ID: (Human) Q16610, (Mouse) Q61508, (Rat) Q62894
Entrez Gene ID: (Bovine) 524222, (Dog) 608791, (Human) 1893, (Pig) 100620701, (Horse) 100054714, (Rabbit) 100338579, (Guinea pig) 100721378, (Mouse) 13601, (Rat) 116662
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.