Proteintech
Manufacturer Code:115211AP150UL
Catalog # 50-172-7232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ECM1 Fusion Protein Ag2084 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECM1 extracellular matrix protein 1 Secretory component p85; Extracellular matrix protein 1; Secretory component p85; testicular tissue protein Li 61
Gene Aliases: AI663821; ECM1; p85; URBWD
UniProt ID: (Human) Q16610, (Mouse) Q61508, (Rat) Q62894
Entrez Gene ID: (Human) 1893, (Mouse) 13601, (Rat) 116662
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.