Proteintech
Manufacturer Code:661171IG150UL
Catalog # 50-173-6553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B9D5 |
Immunogen | ECHS1 Fusion Protein Ag16775 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ECHS1 Enoyl CoA hydratase 1 SCEH; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; Enoyl-CoA hydratase 1; Enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; SCEH; short chain enoyl-CoA hydratase; Short-chain enoyl-CoA hydratase
Gene Aliases: ECHS1; ECHS1D; SCEH
UniProt ID: (Human) P30084
Entrez Gene ID: (Human) 1892
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.