Invitrogen
Manufacturer Code:PA599975
Catalog # PIPA599975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ECHS1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C80529 Echs1 ECHS1D enoyl CoA hydratase short chain 1 mitochondrial enoyl Coenzyme A hydratase short chain 1 enoyl Coenzyme A hydratase short chain 1 mitochondrial enoyl-CoA hydratase 1 Enoyl-CoA hydratase short chain 1 mitochondrial Enoyl-CoA hydratase mitochondrial enoyl-CoA hydratase short chain 1 Enoyl-CoA hydratase short chain 1 mitochondrial enoyl-CoA hydratase short chain 1 mitochondrial mitochondrial short-chain enoyl-coenzyme A hydratase 1 SCEH short chain enoyl-CoA hydratase short-chain enoyl-CoA hydratase; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain 1; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; Enoyl-CoA hydratase 1; Enoyl-CoA hydratase short chain 1 mitochondrial; Enoyl-CoA hydratase, mitochondrial; Enoyl-CoA hydratase, short chain 1, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; mitochondrial short-chain enoyl-coenzyme A hydratase 1; SCEH; short chain enoyl-CoA hydratase; Short-chain enoyl-CoA hydratase
Gene Aliases: C80529; ECHS1; ECHS1D; SCEH
UniProt ID: (Human) P30084, (Rat) P14604, (Mouse) Q8BH95
Entrez Gene ID: (Human) 1892, (Rat) 140547, (Mouse) 93747
Molecular Function: acetyltransferase acyltransferase dehydrogenase epimerase/racemase hydratase isomerase ligase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.