Invitrogen
Manufacturer Code:PA530013
Catalog # PIPA530013
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 55 and 290 of Human ECHS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; Enoyl-CoA hydratase 1; Enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; SCEH; SCEH enoyl Coenzyme A hydratase short chain 1 mitochondrial enoyl-CoA hydratase 1 enoyl-CoA hydratase mitochondrial short-chain enoyl-CoA hydratase; short chain enoyl-CoA hydratase; Short-chain enoyl-CoA hydratase
Gene Aliases: C80529; ECHS1; ECHS1D; SCEH
UniProt ID: (Human) P30084, (Mouse) Q8BH95
Entrez Gene ID: (Human) 1892, (Mouse) 93747
Molecular Function: acetyltransferase acyltransferase dehydrogenase epimerase/racemase hydratase isomerase ligase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.