Novus Biologicals
Manufacturer Code:NBP296977IR
Catalog # NB041530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ECHS1 (AAH08906.1 28A-290Q). |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 4.2.1 enoyl CoA hydratase short chain 1 mitochondrial enoyl Coenzyme A hydratase short chain 1 mitochondrial Enoyl-CoA hydratase 1 enoyl-CoA hydratase mitochondrial SCEHEC 4.2.1.17 Short-chain enoyl-CoA hydratase; enoyl CoA hydratase, short chain, 1, mitochondrial; enoyl Coenzyme A hydratase, short chain, 1, mitochondrial; Enoyl-CoA hydratase 1; Enoyl-CoA hydratase, mitochondrial; enoyl-CoA hydratase, short chain, 1, mitochondrial; SCEH; short chain enoyl-CoA hydratase; Short-chain enoyl-CoA hydratase
Gene Aliases: ECHS1; ECHS1D; SCEH
UniProt ID: (Human) P30084
Entrez Gene ID: (Human) 1892
Molecular Function: acetyltransferase acyltransferase dehydrogenase epimerase/racemase hydratase isomerase ligase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.