Invitrogen
Manufacturer Code:PA5111064
Catalog # PIPA5111064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human ECHDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700028A24Rik AI314462 AI930038 BOS_9528 D10Ertd667e dJ351K20.2 Echdc1 Enoyl-CoA hydratase domain-containing protein 1 Ethylmalonyl-CoA decarboxylase HEL-S-76 Methylmalonyl-CoA decarboxylase MMCD; enoyl CoA hydratase domain containing 1; enoyl Coenzyme A hydratase domain containing 1; Enoyl-CoA hydratase domain-containing protein 1; epididymis secretory protein Li 76; Ethylmalonyl-CoA decarboxylase; Methylmalonyl-CoA decarboxylase; MMCD
Gene Aliases: dJ351K20.2; ECHDC1; HEL-S-76; MMCD
UniProt ID: (Human) Q9NTX5
Entrez Gene ID: (Human) 55862
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.