Invitrogen
Manufacturer Code:PA537009
Catalog # PIPA537009
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 518-563 of human EHHADH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3,2-trans-enoyl-CoA isomerase; 3-hydroxyacyl-CoA dehydrogenase; Enoyl-CoA hydratase/3,2-trans-enoyl-CoA isomerase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-bifunctional enzyme; L-bifunctional protein; L-bifunctional protein, peroxisomal; L-PBE; L-peroxisomal bifunctional enzyme; L-specific bifunctional protein; L-specific multifunctional beta-oxdiation protein; LBP; MFE1; MFP-1; MFP1; Multifunctional enzyme 1; multifunctional enzyme type 1; Multifunctional protein 1; PBE; Peroxisomal bifunctional enzyme; Peroxisomal bifunctional enzyme PBE PBFE Enoyl-CoA hydratase/32-trans-enoyl-CoA isomerase 3-hydroxyacyl-CoA dehydrogenase ECHD; peroxisomal bifunctional enzyme type 1; peroxisomal enoyl-CoA hydratase
Gene Aliases: ECHD; EHHADH; HD; LBFP; LBP; MEF; Mfe; MFP; Pbe; PBFE; 1; 1300002P22Rik; FRTS3; L-PBE; Mfe1; MFP1; perMFE-1
UniProt ID: (Human) Q08426, (Mouse) Q9DBM2, (Rat) P07896
Entrez Gene ID: (Human) 1962, (Mouse) 74147, (Rat) 171142
Molecular Function:
dehydrogenase
epimerase/racemase
hydratase
isomerase
lyase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.