Novus Biologicals
Manufacturer Code:NBP183215
Catalog # NBP183215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:EVDVGLAADVGTLQRLPKVIGNQSLVNELAFTARKMMADEALGSGLVSRVFPDKEVMLDAALALAAEISSKSPVAVQSTKVNLLYSRDHSVAESLNYVASWNMSMLQTQDLVKSVQATTENKELKTVTF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial; delta(35)-Delta(24)-dienoyl-CoA isomerase mitochondrial delta35-delta24-dienoyl-CoA isomerase dienoyl-CoA isomerase EC 5.3.3.- enoyl CoA hydratase 1 peroxisomal enoyl Coenzyme A hydratase 1 peroxisomal HPXEL peroxisomal enoyl-CoA hydratase 1; delta3,5-delta2,4-dienoyl-CoA isomerase; delta3,5-delta2,4-dienoyl-coenzyme A isomerase; dienoyl-CoA isomerase; enoyl CoA hydratase 1, peroxisomal; enoyl Coenzyme A hydratase 1, peroxisomal; enoyl-CoA hydratase 1, peroxisomal; peroxisomal enoyl-CoA hydratase 1
Gene Aliases: ECH1; HPXEL
UniProt ID: (Human) Q96EZ9
Entrez Gene ID: (Human) 1891
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.