Invitrogen
Manufacturer Code:MA523941
Catalog # PIMA523941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 233914 |
Immunogen | Mouse myeloma cell line NS0-derived recombit human ECE-2 Val104-Leu787 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: ECE-2; ECE2KIAA0604 Endothelin-converting enzyme 2 Methyltransferase-like region Endothelin-converting enzyme 2 region; EEF1AKMT4-ECE2 readthrough transcript protein; Endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; Methyltransferase-like region
Gene Aliases: ECE2; EEF1AKMT4-ECE2; KIAA0604; UNQ403/PRO740
UniProt ID: (Human) P0DPD8
Entrez Gene ID: (Human) 9718
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.