Invitrogen
Manufacturer Code:PA569351
Catalog # PIPA569351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat ECE2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECE-2; ECE-2a-1; ECE-2a-2; ECE-2b-1; ECE-2b-2; ECE2 KIAA0604 Endothelin-converting enzyme 2 Methyltransferase-like region Endothelin-converting enzyme 2 region; EEF1A lysine methyltransferase 4; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; Endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; Methyltransferase-like region
Gene Aliases: 1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; ECE2; EEF1AKMT4; EEF1AKMT4-ECE2; KIAA0604; UNQ403/PRO740
UniProt ID: (Human) P0DPD8, (Bovine) P0DPE2, (Mouse) B2RQR8
Entrez Gene ID: (Rabbit) 100348333, (Human) 9718, (Horse) 100059042, (Bovine) 281134, (Dog) 488105, (Guinea pig) 100725061, (Mouse) 107522, (Rat) 408243
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.