Invitrogen
Manufacturer Code:PA5100495
Catalog # PIPA5100495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ECE2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810009K13Rik 6330509A19Rik 9630025D12Rik BB127715 Ece2 ECE-2 EEF1AKMT4-ECE2 EEF1AKMT4-ECE2 readthrough transcript protein endothelin converting enzyme 2 endothelin-converting enzyme 2 Endothelin-converting enzyme 2 region KIAA0604 Methyltransferase-like region UNQ403/PRO740; ECE-2; EEF1AKMT4-ECE2 readthrough transcript protein; endothelin converting enzyme 2; Endothelin-converting enzyme 2; Endothelin-converting enzyme 2 region; Methyltransferase-like region
Gene Aliases: 1810009K13Rik; 6330509A19Rik; 9630025D12Rik; BB127715; ECE2; EEF1AKMT4-ECE2; KIAA0604; UNQ403/PRO740
UniProt ID: (Human) P0DPD8, (Mouse) B2RQR8
Entrez Gene ID: (Human) 9718, (Mouse) 107522, (Rat) 408243
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.