Invitrogen
Manufacturer Code:PA5102821
Catalog # PIPA5102821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ECE1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW322500 Ece ECE 1 ECE1 ECE-1 ECE-1a ECE-1b ECE-1d endothelin converting enzyme 1 Endothelin-converting enzyme 1 endothelin-converting enzyme-1a endothelin-converting enzyme-1b endothelin-converting enzyme-1d; ECE-1; Endothelin-converting enzyme 1; endothelin-converting enzyme-1a; endothelin-converting enzyme-1b; endothelin-converting enzyme-1d
Gene Aliases: AW322500; ECE; ECE-1; ECE-1a; ECE-1b; ECE-1d; ECE1
UniProt ID: (Human) P42892, (Mouse) Q4PZA2
Entrez Gene ID: (Human) 1889, (Rat) 94204, (Mouse) 230857
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.