Novus Biologicals
Manufacturer Code:NBP270612IR
Catalog # NBP270612IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B6 |
Immunogen | Recombinant protein fragment corresponding to amino acids 486-770 of human ECE-1 (NP_001388) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.4.24 EC 3.4.24.71 ECE ECE-1 endothelin converting enzyme 1 endothelin-converting enzyme 1; ECE-1; Endothelin-converting enzyme 1
Gene Aliases: ECE; ECE1
UniProt ID: (Human) P42892
Entrez Gene ID: (Human) 1889
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.