R&D Systems
Manufacturer Code:MAB17841
Catalog # MAB17841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 303913 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human ECE-1 Gln90-Trp770 Accession # P42892 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.4.24 EC 3.4.24.71 ECE ECE1 ECE-1 endothelin converting enzyme 1 endothelin-converting enzyme 1; ECE-1; Endothelin-converting enzyme 1
Gene Aliases: ECE; ECE1
UniProt ID: (Human) P42892
Entrez Gene ID: (Human) 1889
Molecular Function:
hydrolase
metalloprotease
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.