Novus Biologicals
Manufacturer Code:NBP2829710.1ML
Catalog # NB030900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human EBPL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBRPEmopamil-binding-related protein emopamil binding protein-like emopamil binding related protein delta8-delta7 sterol isomerase relatedprotein emopamil-binding protein-like ERP; emopamil binding protein-like; emopamil binding related protein, delta8-delta7 sterol isomerase related protein; Emopamil-binding protein-like; Emopamil-binding-related protein
Gene Aliases: EBPL; EBRP; ERP
UniProt ID: (Human) Q9BY08
Entrez Gene ID: (Human) 84650
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.