Invitrogen
Manufacturer Code:MA525620
Catalog # PIMA525620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D3 |
Immunogen | Full length human recombit protein of PA2G4 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 38kDaAA672939Cell cycle protein p38 2G4 homologCell cycle protein p38-2G4 homologErbB-3 binding protein 1ErbB3 binding protein 1ErbB3-binding protein 1ErbB3-binding protein Ebp1hG4 1hG4-1IRES-specific cellular trans-acting factor 45 kDaMGC81621MGC94070Mpp1p38 2G4PA2G4PA2G4&lowbarHUMANPlfapProliferation associated 2G4Proliferation associated 2G4 38-KDProliferation-associated 2G4 38kDaProliferation-associated 2G4 aProliferation-associated protein 1Proliferation-associated protein 2G4Protein p38-2G4si:dz150i12.2wu:fb19b11wu:ft56d05zgc:86732; Cell cycle protein p38-2G4 homolog; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4-1; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; Proliferation-associated protein 2G4
Gene Aliases: EBP1; HG4-1; p38-2G4; PA2G4
UniProt ID: (Human) Q9UQ80
Entrez Gene ID: (Human) 5036
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.