Invitrogen
Manufacturer Code:MA535648
Catalog # PIMA535648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1281 |
Immunogen | A synthesized peptide derived from human EBP1 / PA2G4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 38kDa AA672939 Cell cycle protein p38-2G4 homolog Ebp1 ErbB3-binding protein 1 hG4 1 hG4-1 IRES-specific cellular trans-acting factor 45 kDa ITAF45 MGC81621 MGC94070 Mpp1 p38 2G4 p38-2G4 PA2G4 Plfap Proliferation-associated protein 1 Proliferation-associated protein 2G4 Protein p38-2G4 si:dz150i12.2 wu:fb19b11 wu:ft56d05 zgc:86732; Cell cycle protein p38-2G4 homolog; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4-1; IRES-specific cellular trans-acting factor 45 kDa; ITAF45; Mpp1; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; Proliferation-associated protein 1; Proliferation-associated protein 2G4; Protein p38-2G4
Gene Aliases: 38kDa; AA672939; EBP1; HG4-1; p38-2G4; PA2G4; Plfap
UniProt ID: (Human) Q9UQ80, (Rat) Q6AYD3, (Mouse) P50580
Entrez Gene ID: (Human) 5036, (Rat) 288778, (Mouse) 18813
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.