Invitrogen
Manufacturer Code:PA512467
Catalog # PIPA512467
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 228-255 amino acids from the central region of human EBP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle protein p38-2G4 homolog; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4-1; IRES-specific cellular trans-acting factor 45 kDa; ITAF45; Mpp1; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; Proliferation-associated protein 1; Proliferation-associated protein 2G4; Proliferation-associated protein 2G4 Cell cycle protein p38-2G4 homolog hG4-1 ErbB3-binding protein 1 PA2G4 EBP1; Protein p38-2G4
Gene Aliases: 38kDa; AA672939; EBP1; HG4-1; p38-2G4; PA2G4; Plfap
UniProt ID: (Human) Q9UQ80, (Mouse) P50580
Entrez Gene ID: (Human) 5036, (Mouse) 18813
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.