Novus Biologicals
Manufacturer Code:NBP2829650.1ML
Catalog # NB030894
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of EBNA1BP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBNA1 binding protein 2 EBNA1-binding protein 2nucleolar protein p40 homolog of yeast EBNA1-binding protein EBP2nuclear FGF3 binding protein NOBP Nucleolar protein p40 P40 probable rRNA-processing protein EBP2; EBNA1-binding protein 2; nuclear FGF3 binding protein; Nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; Probable rRNA-processing protein EBP2
Gene Aliases: EBNA1BP2; EBP2; NOBP; P40
UniProt ID: (Human) Q99848
Entrez Gene ID: (Human) 10969
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.