Invitrogen
Manufacturer Code:PA523228
Catalog # PIPA523228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (73 RGHSWPCLQQTPTSTSC 89) of EBI3 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cytokine receptor; EBV-induced gene 3 protein; Epstein-Barr virus induced gene 3; Epstein-Barr virus-induced gene 3 protein; Epstein-Barr virus-induced gene 3 protein homolog; IL-27 subunit beta; IL-27B; IL-27B IL27B EBV-induced gene 3 protein Epstein-Barr virus induced gene 3 IL-27 subunit beta IL27 subunit IL35 subunit cytokine receptor epstein-Barr virus-induced gene 3 protein interleukin-27 subunit beta; IL27 subunit; IL35 subunit; Interleukin-27 subunit beta
Gene Aliases: EBI-3; EBI3; IL-27; IL-27B; IL27B
UniProt ID: (Human) Q14213, (Mouse) O35228
Entrez Gene ID: (Human) 10148, (Mouse) 50498
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.