R&D Systems
Manufacturer Code:MAB10272SP
Catalog # MAB10272SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 486633 |
Immunogen | Mouse myeloma cell line NS0-derived transfected withrecombinant human EBI2 Accession # P32249 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: EBI2; EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor) EBV-induced G-protein coupled receptor 2 Epstein-Barr virus-induced G-protein coupled receptor 2 G protein-coupled receptor 183 GPR183 G-protein coupled receptor 183; EBV-induced G-protein coupled receptor 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); Epstein-Barr virus-induced G-protein coupled receptor 2; G-protein coupled receptor 183
Gene Aliases: EBI2; GPR183
UniProt ID: (Human) P32249
Entrez Gene ID: (Human) 1880
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.