Novus Biologicals
Manufacturer Code:NBP2866260.1ML
Catalog # NB034250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human EBI2/GPR183 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EBI2; EBI2Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupledreceptor) EBV-induced G-protein coupled receptor 2 Epstein-Barr virus-induced G-protein coupled receptor 2 G protein-coupled receptor 183 G-protein coupled receptor 183; EBV-induced G-protein coupled receptor 2; Epstein-Barr virus induced gene 2 (lymphocyte-specific G protein-coupled receptor); Epstein-Barr virus-induced G-protein coupled receptor 2; G-protein coupled receptor 183
Gene Aliases: EBI2; GPR183
UniProt ID: (Human) P32249
Entrez Gene ID: (Human) 1880
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.