Invitrogen
Manufacturer Code:PA5113949
Catalog # PIPA5113949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C terminal region of human EBF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COE4 early B cell factor 4 early B-cell factor 3 Early B-cell factor 4 early B-cell factor 4-like EBF family member 4 Ebf3 EBF4 EBF-4 KIAA1442 O/E-4 OE-4 Olf-1 Olf-1/EBF-like 4 RP5-860F19.3 Transcription factor COE4; Early B-cell factor 4; EBF-4; O/E-4; OE-4; Olf-1/EBF-like 4; RP5-860F19.3; Transcription factor COE4
Gene Aliases: COE4; EBF4; KIAA1442; O/E-4
UniProt ID: (Human) Q9BQW3
Entrez Gene ID: (Human) 57593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.