Invitrogen
Manufacturer Code:PA518720
Catalog # PIPA518720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (HNQLPALANTSV) corresponding to the internal amino acids of EBF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Collier, Olf and EBF transcription factor 1; Early B-cell factor; EBF1 early B-cell factor 1 COE1 EBF O/E-1 OLF1 Collier Olf and EBF transcription factor 1 early B-cell factor olfactory neuronal transcription factor 1; O/E-1; olfactory neuronal transcription factor 1; Transcription factor COE1
Gene Aliases: COE1; EBF; EBF1; O/E-1; OE-1; Olf-1; OLF1
UniProt ID: (Human) Q9UH73, (Mouse) Q07802
Entrez Gene ID: (Human) 1879, (Mouse) 13591
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.