Novus Biologicals
Manufacturer Code:NBP147292
Catalog # NBP147292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 163 and 213 of human Receptor-binding Cancer Antigen Expressed on SiSo cells using the numbering given in entry NP_004206.1 (GeneID 9166). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer-associated surface antigen RCAS1; Cancer-associated surface antigen RCAS1 EB9cancer associated surface antigen estrogen receptor binding site associated antigen 9 Estrogen receptor-binding fragment-associated gene 9 protein RCAS1PDAF receptor-binding cancer antigen expressed on SiSo cells; Estrogen receptor-binding fragment-associated gene 9 protein; Receptor-binding cancer antigen expressed on SiSo cells
Gene Aliases: EB9; EBAG9; PDAF; RCAS1
UniProt ID: (Human) O00559
Entrez Gene ID: (Human) 9166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.