Invitrogen
Manufacturer Code:PA579183
Catalog # PIPA579183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human EBAG9 recombit protein (Position: R31-S213). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer-associated surface antigen RCAS1; Estrogen receptor-binding fragment-associated gene 9 protein; Receptor-binding cancer antigen expressed on SiSo cells; Receptor-binding cancer antigen expressed on SiSo cells Cancer-associated surface antigen RCAS1 Estrogen receptor-binding fragment-associated gene 9 protein EBAG9 RCAS1
Gene Aliases: AI835379; EB9; EBAG9; PDAF; RCAS1
UniProt ID: (Human) O00559, (Mouse) Q9D0V7, (Rat) Q5PQP2
Entrez Gene ID: (Human) 9166, (Mouse) 55960, (Rat) 299864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.