Abnova
Manufacturer Code:PAB12726
Catalog # 89-116-056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human EBAG9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C or lower.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of EBAG9.
For Research Use Only
Protein Aliases: Cancer-associated surface antigen RCAS1; EB9; Estrogen receptor-binding fragment-associated gene 9 protein; PDAF; RCAS1; Receptor-binding cancer antigen expressed on SiSo cells
Gene Aliases: EB9; EBAG9; PDAF; RCAS1
UniProt ID: (Human) O00559
Entrez Gene ID: (Human) 9166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.