Novus Biologicals
Manufacturer Code:NBP146151
Catalog # NBP146151
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence SGRLSNVAPPCILR corresponding to internal region according to NP_036458.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: APC binding protein; EB1 protein family member 3; EB1 protein family member 3 EB3EBF3-S End-binding protein 3 microtubule-associated protein RP/EB family member 3 microtubule-associated protein RP/EB family member 3 RP3EBF3APC binding protein; EB3; EBF3; End-binding protein 3; Microtubule-associated protein RP/EB family member 3; microtubule-associated protein, RP/EB family, member 3; RP3
Gene Aliases: EB3; EBF3; EBF3-S; MAPRE3; RP3
UniProt ID: (Human) Q9UPY8
Entrez Gene ID: (Human) 22924
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.