Novus Biologicals
Manufacturer Code:NBP191856
Catalog # NBP191856
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LLDLEKLPEFNRLHLQRLVSNESQRRQLVGKLQVLVEEAFGCQLQNRDVLNPVYVERILLLRQGHICRLQDLVSPVYSYLWTRPAVGRAQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.1.1.17 GluRS Glutamate--tRNA ligase glutamyl-tRNA synthetase 2 mitochondrial (putative) KIAA1970glutamate tRNA ligase 2 mitochondrial MSE1 probable glutamyl-tRNA synthetase mitochondrial; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial
Gene Aliases: COXPD12; EARS2; gluRS; KIAA1970; MSE1
UniProt ID: (Human) Q5JPH6
Entrez Gene ID: (Human) 124454
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.