Abnova
Manufacturer Code:PAB28109
Catalog # 89-129-614
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of recombinant EARS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant EARS2.
For Research Use Only
Protein Aliases: GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; KIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial
Gene Aliases: COXPD12; EARS2; gluRS; KIAA1970; MSE1
UniProt ID: (Human) Q5JPH6
Entrez Gene ID: (Human) 124454
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.