Invitrogen
Manufacturer Code:PA540389
Catalog # PIPA540389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human EAP30 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EAP30 subunit of ELL complex; ELL-associated protein of 30 kDa; ESCRT-II complex subunit VPS22; hVps22; SNF8 ESCRT-II complex subunit homolog (S. cerevisiae) Dot3 EAP30 VPS22; SNF8, ESCRT-II complex subunit; SNF8, ESCRT-II complex subunit, homolog; vacuolar sorting snf8 ELL associated of 30kDa; Vacuolar-sorting protein SNF8
Gene Aliases: D11Moh34; Dot3; EAP30; RGD1310144; si:dkey-220f10.1; SNF8; VPS22; zgc:101578
UniProt ID: (Human) Q96H20, (Mouse) Q9CZ28, (Rat) Q5RK19, (Zebrafish) A4VAK8
Entrez Gene ID: (Dog) 491063, (Rabbit) 100345109, (Human) 11267, (Bovine) 505628, (Horse) 100055830, (Mouse) 27681, (Rat) 287645, (Guinea pig) 100714288, (Zebrafish) 492771
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.