Invitrogen
Manufacturer Code:PA5116085
Catalog # PIPA5116085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TTRAP corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'-Tyr-DNA phosphodiesterase; 5'-Tyr-DNA phosphodiesterase 5'-tyrosyl-DNA phosphodiesterase AD022 AD-022 D13Ertd656e dJ30M3.3 EAP2 EAPII ETS1-associated protein 2 ETS1-associated protein II hTDP2 MGC111021 MGC9099 RP1-30M3.3 Tdp2 Traf and Tnf receptor associated protein TRAF and TNF receptor-associated protein TTRAP Tyr-DNA phosphodiesterase 2 tyrosyl-DNA phosphodiesterase 2 Tyrosyl-RNA phosphodiesterase VPg unlinkase; 5'-tyrosyl-DNA phosphodiesterase; EAPII; ETS1-associated protein 2; ETS1-associated protein II; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; Tyr-DNA phosphodiesterase 2; Tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Aliases: AD-022; AD022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; hTDP2; TDP2; TTRAP
UniProt ID: (Human) O95551, (Mouse) Q9JJX7, (Rat) Q3T1H5
Entrez Gene ID: (Human) 51567, (Mouse) 56196, (Rat) 498749
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.