Invitrogen
Manufacturer Code:MA524498
Catalog # PIMA524498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G2 |
Immunogen | EAF1 (AAH41329 1 a.a. &bsim 268 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL-associated factor 1; ELL-associated factor 1 EAF1
Gene Aliases: EAF1
UniProt ID: (Human) Q96JC9
Entrez Gene ID: (Human) 85403
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.