Novus Biologicals
Manufacturer Code:NBP292383100UL
Catalog # NB026647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 50-120 of human EAF1 (NP_149074.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1 ELL associated factor 1 ELL-associated factor 1; ELL-associated factor 1
Gene Aliases: EAF1
UniProt ID: (Human) Q96JC9
Entrez Gene ID: (Human) 85403
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.