Invitrogen
Manufacturer Code:PA5115152
Catalog # PIPA5115152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EAAT3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D130048G10Rik DCBXA EAAC1 EAAC2 EAAT3 excitatory amino acid carrier 1 excitatory amino acid carrier 2 excitatory amino acid transporter 3 excitatory amino acid transporter-3 excitatory amino-acid carrier 1 glutamate transporter mEAAC2 MEAAC1 neuronal and epithelial glutamate transporter REAAC1 SCZD18 SLC1A1 sodium-dependent glutamate/aspartate transporter 3 solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter system Xag) member 1 Solute carrier family 1 A1 (brain glutamate transporter) solute carrier family 1 member 1 solute carrier family 1 member 1; excitatory amino acid carrier 1; excitatory amino acid carrier 2; Excitatory amino acid transporter 3; Excitatory amino-acid carrier 1; glutamate transporter mEAAC2; Neuronal and epithelial glutamate transporter; Sodium-dependent glutamate/aspartate transporter 3; solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1; Solute carrier family 1 member 1; solute carrier family 1, member 1
Gene Aliases: D130048G10Rik; DCBXA; EAAC1; EAAC2; EAAT3; MEAAC1; SCZD18; SLC1A1
UniProt ID: (Human) P43005, (Mouse) P51906
Entrez Gene ID: (Human) 6505, (Mouse) 20510
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.