Abnova
Manufacturer Code:PAB27527
Catalog # 89-129-036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 480-493 at internal region of Drosophila E2f. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of E2f.
For Research Use Only
Protein Aliases: CG6376; CG6376-PA; CG6376-PB; CG6376-PC; CG6376-PD; CG6376-PE; CG6376-PF; dE2F; dE2F1; def21; DmE2F-1; drosE2F1; DRTF1/E2F; E(Sev-CycE)3A; E(var)3-93E; E(var)3-95E; E(var)93E; E2-promoter binding factor; E2F transcription factor; E2F-1; E2F1; E2f1-PA; E2f1-PB; E2f1-PC; E2f1-PD; E2f1-PE; E2f1-PF; Evar(3)164; l(3)07172; l(3)j3B1; l(3)j3C2; l(3)rM729; LD35741p; Transcription factor E2f1
Gene Aliases: CG6376; dE2F; dE2F1; def21; DmE2F-1; Dmel\CG6376; Dmel_CG6376; Dp; drosE2F1; DRTF1/E2F; E(Sev-CycE)3A; E(var)3-93E; E(var)3-95E; E(var)93E; E2F; E2F-1; E2F1; Evar(3)164; l(3)07172; l(3)j3B1; l(3)j3C2; l(3)rM729
UniProt ID: (Fruit fly) Q27368
Entrez Gene ID: (Fruit fly) 42550
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.