Invitrogen
Manufacturer Code:PA543088
Catalog # PIPA543088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human E2F8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2F family member 8; E2F-8; Transcription factor E2F8
Gene Aliases: 4432406C08Rik; AA410048; E2F-8; E2F8; RGD1308091
UniProt ID: (Human) A0AVK6, (Bovine) E1BKK0, (Mouse) Q58FA4
Entrez Gene ID: (Human) 79733, (Bovine) 786629, (Horse) 100071962, (Dog) 476885, (Rabbit) 100350717, (Guinea pig) 100723781, (Mouse) 108961, (Rat) 308607
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.