Invitrogen
Manufacturer Code:PA5106867
Catalog # PIPA5106867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human E2F8 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4432406C08Rik AA410048 E2F 8 E2F family member 8 E2F transcription factor 8 E2f8 E2F-8 FLJ23311 RGD1308091 Transcription factor E2F8; E2F family member 8; E2F-8; Transcription factor E2F8
Gene Aliases: 4432406C08Rik; AA410048; E2F-8; E2F8; RGD1308091
UniProt ID: (Human) A0AVK6, (Mouse) Q58FA4
Entrez Gene ID: (Human) 79733, (Mouse) 108961, (Rat) 308607
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.