Invitrogen
Manufacturer Code:PA568912
Catalog # PIPA568912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N terminal region of mouse E2F7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2F-7; E2F7 E2F transcription factor 7 FLJ12981; Transcription factor E2F7
Gene Aliases: A630014C11Rik; D10Ertd739e; E2F-7; E2F7
UniProt ID: (Human) Q96AV8, (Mouse) Q6S7F2, (Rat) D4A4D7
Entrez Gene ID: (Human) 144455, (Horse) 100050373, (Dog) 482575, (Rabbit) 100357309, (Mouse) 52679, (Guinea pig) 100734186, (Rat) 314818
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.