Invitrogen
Manufacturer Code:PA570534
Catalog # PIPA570534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat E2F6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2F-6; E2F-binding site-modulating activity protein; E2F6 E2F transcription factor 6 E2F-6 MGC111545 E2F transcription factor 6 isoform 1; EMA; Transcription factor E2F6; transcriptional repressor E2F6
Gene Aliases: AI462434; E2F-6; E2F6; E2F6b; EMA
UniProt ID: (Human) O75461, (Bovine) Q08DY6, (Mouse) O54917
Entrez Gene ID: (Dog) 609996, (Human) 1876, (Horse) 100072285, (Rabbit) 100347358, (Pig) 100737827, (Bovine) 514085, (Guinea pig) 100713523, (Mouse) 50496, (Rat) 313978
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.