Invitrogen
Manufacturer Code:PA579179
Catalog # PIPA579179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human E2F2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2F transcription factor 2; E2F-2; Transcription factor E2F2; Transcription factor E2F2 E2F-2 E2F2
Gene Aliases: E2F-2; E2F2
UniProt ID: (Human) Q14209, (Mouse) P56931
Entrez Gene ID: (Human) 1870, (Mouse) 242705, (Rat) 684111
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.