Novus Biologicals
Manufacturer Code:NB600210AF350
Catalog # NB60210A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 16G7 |
Immunogen | The region encoding the DNA binding domain (amino acids 244-264) of E2F-1 expressed in E. coli. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: E2F transcription factor 1 E2F-1 PBR3 PRB-binding protein E2F-1 RBAP1 RBAP-1 RBBP3 RBBP-3 RBP3 Retinoblastoma-associated protein 1 Retinoblastoma-binding protein 3 transcription factor E2F1; E2F-1; PBR3; PRB-binding protein E2F-1; RBAP-1; RBBP-3; Retinoblastoma-associated protein 1; Retinoblastoma-binding protein 3; Transcription factor E2F1
Gene Aliases: E2F-1; E2F1; RBAP1; RBBP3; RBP3
UniProt ID: (Human) Q01094
Entrez Gene ID: (Human) 1869
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.