R&D Systems
Manufacturer Code:AF4825
Catalog # AF4825
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human E2F-1 Val264-Phe437 Accession # Q01094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: E2F transcription factor 1 E2F1 E2F-1 PBR3 PRB-binding protein E2F-1 RBAP1 RBAP-1 RBBP3 RBBP-3 RBP3 Retinoblastoma-associated protein 1 Retinoblastoma-binding protein 3 transcription factor E2F1; E2F-1; PBR3; PRB-binding protein E2F-1; RBAP-1; RBBP-3; Retinoblastoma-associated protein 1; Retinoblastoma-binding protein 3; Transcription factor E2F1
Gene Aliases: E2F-1; E2F1; RBAP1; RBBP3; RBP3
UniProt ID: (Human) Q01094
Entrez Gene ID: (Human) 1869
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.