Novus Biologicals
Manufacturer Code:NBP143393APC
Catalog # NBP143393AJ
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | P2H3 |
Immunogen | The immunogen for this antibody was CD62E. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD62 antigen-like family member E; CD62 antigen-like family member E CD62E CD62E antigen ELAM ELAM-1 ELAM1E-selectin endothelial adhesion molecule 1 Endothelial leukocyte adhesion molecule 1 ESEL LECAM2 leukocyte endothelial cell adhesion molecule 2 Leukocyte-endothelial cell adhesion molecule 2 selectin E; CD62E; E-selectin; ELAM-1; endothelial adhesion molecule 1; Endothelial leukocyte adhesion molecule 1; LECAM2; leukocyte endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 2
Gene Aliases: CD62E; ELAM; ELAM1; ESEL; LECAM2; SELE
UniProt ID: (Human) P16581
Entrez Gene ID: (Human) 6401
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.